Gene Transcripts
- Gene Symbol
- THEG5
- Entrez Gene ID
- 100507527
- Gene Name
- testis highly expressed protein 5
- Gene Aliases
- Chromosome Location
- Chr. 19: 31573343 - 31593550 on Build GRCh38
- UniGene ID
- Hs.432564
- Species
- Homo sapiens
- Gene Symbol
- LOC101927277
- Entrez Gene ID
- 101927277
- Gene Name
- uncharacterized LOC101927277
- Gene Aliases
- Chromosome Location
- Chr. 19: 31588040 - 31595720 on Build GRCh38
- UniGene ID
- Hs.707861
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CR627061.1 |
- |
2 |
899 |
|
View Full Product Details