Gene Transcripts

Gene Symbol
H1FX-AS1
Entrez Gene ID
339942
Gene Name
H1FX antisense RNA 1
Gene Aliases
C3orf47
Chromosome Location
Chr. 3: 129316271 - 129324569 on Build GRCh38
UniGene ID
Hs.450096
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026991.1 3, 4 1545
GenBank mRNA AL040547.1 - 2, 3 636
BC093941.1 - 2, 3 208
BC101739.1 - 2, 3 208

View Full Product Details