Gene Transcripts

Gene Symbol
CXorf66
Entrez Gene ID
347487
Gene Name
chromosome X open reading frame 66
Gene Aliases
SGPX
Chromosome Location
Chr. X: 139955725 - 139965518 on Build GRCh38
UniGene ID
Hs.121384
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013403.2 NP_001013421.1 3 621
GenBank mRNA BC137148.1 - 3 621
BC137149.1 - 3 621 9021146

View Full Product Details