Gene Transcripts
- Gene Symbol
- RPS3A
- Entrez Gene ID
- 6189
- Gene Name
- ribosomal protein S3A
- Gene Aliases
- FTE1, MFTL, S3A
- Chromosome Location
- Chr. 4: 151099573 - 151104652 on Build GRCh38
- UniGene ID
- Hs.356572
- Species
- Homo sapiens
- Gene Symbol
- ATP8B3
- Entrez Gene ID
- 148229
- Gene Name
- ATPase phospholipid transporting 8B3
- Gene Aliases
- ATPIK
- Chromosome Location
- Chr. 19: 1782070 - 1812276 on Build GRCh38
- UniGene ID
- Hs.306212
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK125225.1 |
- |
14 |
2274 |
|
- Gene Symbol
- BMP8B
- Entrez Gene ID
- 656
- Gene Name
- bone morphogenetic protein 8b
- Gene Aliases
- BMP8, OP2
- Chromosome Location
- Chr. 1: 39757182 - 39788861 on Build GRCh38
- UniGene ID
- Hs.734367
- Species
- Homo sapiens
- Gene Symbol
- CARMN
- Entrez Gene ID
- 728264
- Gene Name
- cardiac mesoderm enhancer-associated non-coding RNA
- Gene Aliases
- CARMEN, MIR143HG
- Chromosome Location
- Chr. 5: 149406845 - 149432836 on Build GRCh38
- UniGene ID
- Hs.519666
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
GQ292874.1 |
- |
1 |
527 |
|
- Gene Symbol
- CYP20A1
- Entrez Gene ID
- 57404
- Gene Name
- cytochrome P450 family 20 subfamily A member 1
- Gene Aliases
- CYP-M
- Chromosome Location
- Chr. 2: 203238441 - 203305840 on Build GRCh38
- UniGene ID
- Hs.446065
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_177538.2 |
NP_803882.1
|
13 |
6082 |
|
- Gene Symbol
- IFT81
- Entrez Gene ID
- 28981
- Gene Name
- intraflagellar transport 81
- Gene Aliases
- CDV-1, CDV-1R, CDV1, CDV1R, DV1
- Chromosome Location
- Chr. 12: 110124335 - 110218795 on Build GRCh38
- UniGene ID
- Hs.528382
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK091549.1 |
- |
19 |
1875 |
|
- Gene Symbol
- LINC00926
- Entrez Gene ID
- 283663
- Gene Name
- long intergenic non-protein coding RNA 926
- Gene Aliases
- Chromosome Location
- Chr. 15: 57300365 - 57307769 on Build GRCh38
- UniGene ID
- Hs.181297
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK123700.1 |
- |
1 |
912 |
|
- Gene Symbol
- LOC101929732
- Entrez Gene ID
- 101929732
- Gene Name
- uncharacterized LOC101929732
- Gene Aliases
- LA16c-390H2.4
- Chromosome Location
- Chr. 16: 3542739 - 3559589 on Build GRCh38
- UniGene ID
- Hs.592091
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK125583.1 |
- |
1 |
1198 |
|
- Gene Symbol
- LRRC36
- Entrez Gene ID
- 55282
- Gene Name
- leucine rich repeat containing 36
- Gene Aliases
- RORBP70, XLHSRF2
- Chromosome Location
- Chr. 16: 67326844 - 67385206 on Build GRCh38
- UniGene ID
- Hs.125139
- Species
- Homo sapiens
- Gene Symbol
- MEIOC
- Entrez Gene ID
- 284071
- Gene Name
- meiosis specific with coiled-coil domain
- Gene Aliases
- C17orf104
- Chromosome Location
- Chr. 17: 44656394 - 44677086 on Build GRCh38
- UniGene ID
- Hs.349758
- Species
- Homo sapiens
- Gene Symbol
- PCSK5
- Entrez Gene ID
- 5125
- Gene Name
- proprotein convertase subtilisin/kexin type 5
- Gene Aliases
- PC5, PC6, PC6A, SPC6
- Chromosome Location
- Chr. 9: 75890601 - 76362339 on Build GRCh38
- UniGene ID
- Hs.368542
- Species
- Homo sapiens
- Gene Symbol
- PPIAL4C
- Entrez Gene ID
- 653598
- Gene Name
- peptidylprolyl isomerase A like 4C
- Gene Aliases
- COAS-2
- Chromosome Location
- Chr. 1: 149583823 - 149584607 on Build GRCh38
- UniGene ID
- Hs.534674
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK123006.1 |
- |
2 |
2805 |
|
- Gene Symbol
- PPP1R26
- Entrez Gene ID
- 9858
- Gene Name
- protein phosphatase 1 regulatory subunit 26
- Gene Aliases
- KIAA0649
- Chromosome Location
- Chr. 9: 135477650 - 135488894 on Build GRCh38
- UniGene ID
- Hs.533260
- Species
- Homo sapiens
- Gene Symbol
- SAMM50
- Entrez Gene ID
- 25813
- Gene Name
- SAMM50 sorting and assembly machinery component
- Gene Aliases
- CGI-51, OMP85, SAM50, TOB55, TRG-3, YNL026W
- Chromosome Location
- Chr. 22: 43955381 - 43996532 on Build GRCh38
- UniGene ID
- Hs.505824
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK091282.1 |
- |
1 |
441 |
|
- Gene Symbol
- TMEM132B
- Entrez Gene ID
- 114795
- Gene Name
- transmembrane protein 132B
- Gene Aliases
- Chromosome Location
- Chr. 12: 125186750 - 125662377 on Build GRCh38
- UniGene ID
- Hs.524838
- Species
- Homo sapiens
- Gene Symbol
- TNPO2
- Entrez Gene ID
- 30000
- Gene Name
- transportin 2
- Gene Aliases
- IPO3, KPNB2B, TRN2
- Chromosome Location
- Chr. 19: 12699194 - 12723996 on Build GRCh38
- UniGene ID
- Hs.416049
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK022217.1 |
- |
1 |
1887 |
|
- Gene Symbol
- WDR37
- Entrez Gene ID
- 22884
- Gene Name
- WD repeat domain 37
- Gene Aliases
- Chromosome Location
- Chr. 10: 1056836 - 1132297 on Build GRCh38
- UniGene ID
- Hs.188495
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK128194.1 |
- |
1 |
2111 |
|
- Gene Symbol
- ZNF578
- Entrez Gene ID
- 147660
- Gene Name
- zinc finger protein 578
- Gene Aliases
- Chromosome Location
- Chr. 19: 52453514 - 52516878 on Build GRCh38
- UniGene ID
- Hs.157287
- Species
- Homo sapiens
View Full Product Details