Gene Transcripts

Gene Symbol
TRANK1
Entrez Gene ID
9881
Gene Name
tetratricopeptide repeat and ankyrin repeat containing 1
Gene Aliases
LBA1
Chromosome Location
Chr. 3: 36826817 - 36945662 on Build GRCh38
UniGene ID
Hs.170999
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014831.2 NP_055646.2 2, 3 389
XM_006713432.3 XP_006713495.1 3, 4 557
XM_011534289.2 XP_011532591.1 3, 4 1318
XM_017007570.1 XP_016863059.1 3, 4 360
XM_017007571.1 XP_016863060.1 3, 4 450
XM_017007572.1 XP_016863061.1 3, 4 352
XM_017007573.1 XP_016863062.1 3, 4 360
GenBank mRNA CD698859.1 - 2, 3 348
CN482191.1 - 2, 3 390

View Full Product Details