Gene Transcripts

Gene Symbol
RAB19
Entrez Gene ID
401409
Gene Name
RAB19, member RAS oncogene family
Gene Aliases
RAB19B
Chromosome Location
Chr. 7: 140403740 - 140426881 on Build GRCh38
UniGene ID
Hs.583545
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008749.2 NP_001008749.2 4 747
XM_005249996.3 XP_005250053.1 4 1030
XM_005249997.3 XP_005250054.1 4 1013
XM_005249998.3 XP_005250055.1 4 1008
XM_017012207.1 XP_016867696.1 4 943
GenBank mRNA BC140796.1 - 3 617
BC171745.1 - 3 617

View Full Product Details