Gene Transcripts
- Gene Symbol
- CENPP
- Entrez Gene ID
- 401541
- Gene Name
- centromere protein P
- Gene Aliases
- CENP-P
- Chromosome Location
- Chr. 9: 92325457 - 92615164 on Build GRCh38
- UniGene ID
- Hs.713775
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001012267.2 |
NP_001012267.1
|
6, 7 |
1181 |
|
NM_001286969.1 |
NP_001273898.1
|
5, 6 |
999 |
|
NM_001286971.1 |
NP_001273900.1
|
2, 3 |
338 |
|
GenBank mRNA |
AK091247.1 |
- |
2, 3 |
339 |
|
AK092603.1 |
- |
6, 7 |
996 |
|
AK307262.1 |
- |
2, 3 |
221 |
|
AK307435.1 |
- |
5, 6 |
519 |
|
AL833701.1 |
- |
1, 2 |
856 |
|
BC071726.1 |
- |
6, 7 |
707 |
5554572 |
BX537851.1 |
- |
5, 6 |
528 |
|
View Full Product Details