Gene Transcripts
- Gene Symbol
- SEPT7P9
- Entrez Gene ID
- 285961
- Gene Name
- septin 7 pseudogene 9
- Gene Aliases
- CDC10L, SEPT7L, bA291L22.2
- Chromosome Location
- Chr. 10: 38383069 - 38402852 on Build GRCh38
- UniGene ID
- Hs.406568
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027269.1 |
|
1 |
92 |
|
GenBank mRNA |
BC171738.1 |
- |
1 |
58 |
|
View Full Product Details