Gene Transcripts
- Gene Symbol
- USP6
- Entrez Gene ID
- 9098
- Gene Name
- ubiquitin specific peptidase 6
- Gene Aliases
- HRP1, TRE17, TRE2, TRESMCR, Tre-2, USP6-short
- Chromosome Location
- Chr. 17: 5116438 - 5175032 on Build GRCh38
- UniGene ID
- Hs.448851
- Species
- Homo sapiens
- Gene Symbol
- TBC1D3P5
- Entrez Gene ID
- 440419
- Gene Name
- TBC1 domain family member 3 pseudogene 5
- Gene Aliases
- Chromosome Location
- Chr. 17: 27418005 - 27431623 on Build GRCh38
- UniGene ID
- Hs.385755
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC112354.1 |
- |
1 |
413 |
|
View Full Product Details