Gene Transcripts
- Gene Symbol
- GUSBP4
- Entrez Gene ID
- 375513
- Gene Name
- glucuronidase, beta pseudogene 4
- Gene Aliases
- C6orf216, GUSBL2, SMA3-L, SMAC3L
- Chromosome Location
- Chr. 6: 57908553 - 57936929 on Build GRCh38
- UniGene ID
- Hs.561539
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_132999.1 |
|
5 |
4735 |
|
GenBank mRNA |
AK026691.1 |
- |
1 |
2278 |
|
AL831827.1 |
- |
1 |
5115 |
|
View Full Product Details