Gene Transcripts

Gene Symbol
C5orf56
Entrez Gene ID
441108
Gene Name
chromosome 5 open reading frame 56
Gene Aliases
Chromosome Location
Chr. 5: 132410773 - 132476044 on Build GRCh38
UniGene ID
Hs.658288
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_045116.1 2 353
GenBank mRNA AK096941.1 - 2 145
AK128882.1 - 2 142
AK309191.1 - 2 145
BC130299.1 - 1 43 40146330
BC130656.1 - 1 43 40146687
BM998490.1 - 3 465
DC350817.1 - 2 179
DN991436.1 - 2 361

View Full Product Details