Gene Transcripts
- Gene Symbol
- SLC25A52
- Entrez Gene ID
- 147407
- Gene Name
- solute carrier family 25 member 52
- Gene Aliases
- MCART2
- Chromosome Location
- Chr. 18: 31759696 - 31760880 on Build GRCh38
- UniGene ID
- Hs.567708
- Species
- Homo sapiens
- Gene Symbol
- SLC25A51
- Entrez Gene ID
- 92014
- Gene Name
- solute carrier family 25 member 51
- Gene Aliases
- CG7943, MCART1
- Chromosome Location
- Chr. 9: 37877575 - 37904353 on Build GRCh38
- UniGene ID
- Hs.634120
- Species
- Homo sapiens
- Gene Symbol
- SLC25A51P1
- Entrez Gene ID
- 442229
- Gene Name
- solute carrier family 25 member 51 pseudogene 1
- Gene Aliases
- MCART3P, bA707M13.1
- Chromosome Location
- Chr. 6: 65787879 - 65789483 on Build GRCh38
- UniGene ID
- Hs.390414
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026540.1 |
|
1 |
1037 |
|
GenBank mRNA |
BC024198.1 |
- |
1 |
1038 |
|
View Full Product Details