Gene Transcripts

Gene Symbol
OR2AE1
Entrez Gene ID
81392
Gene Name
olfactory receptor family 2 subfamily AE member 1
Gene Aliases
OR2AE2
Chromosome Location
Chr. 7: 99876062 - 99877033 on Build GRCh38
UniGene ID
Hs.647523
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005276.1 NP_001005276.1 1 827
GenBank mRNA BC137375.1 - 1 851
BC137376.1 - 1 851 9021374

View Full Product Details