Gene Transcripts
- Gene Symbol
- ANKRD30BP2
- Entrez Gene ID
- 149992
- Gene Name
- ankyrin repeat domain 30B pseudogene 2
- Gene Aliases
- C21orf99, CT85, CTSP-1, CTSP1
- Chromosome Location
- Chr. 21: 13038166 - 13118251 on Build GRCh38
- UniGene ID
- Hs.558645
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026916.1 |
|
2 |
304 |
|
- Gene Symbol
- ANKRD30BL
- Entrez Gene ID
- 554226
- Gene Name
- ankyrin repeat domain 30B-like
- Gene Aliases
- ANKRD30BP3, NCRNA00164
- Chromosome Location
- Chr. 2: 132147591 - 132257969 on Build GRCh38
- UniGene ID
- Hs.380689
- Species
- Homo sapiens
View Full Product Details