Gene Transcripts

Gene Symbol
FBXO48
Entrez Gene ID
554251
Gene Name
F-box protein 48
Gene Aliases
Chromosome Location
Chr. 2: 68459419 - 68467325 on Build GRCh38
UniGene ID
Hs.164117
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024680.2 NP_001019851.1 4 826
XM_005264407.2 XP_005264464.1 4 828
XM_017004437.1 XP_016859926.1 4 841
GenBank mRNA BC089423.1 - 4 767
DB303415.1 - 1 385
DQ904332.1 - 4 767

View Full Product Details