Gene Transcripts
- Gene Symbol
- WASH1
- Entrez Gene ID
- 100287171
- Gene Name
- WAS protein family homolog 1
- Gene Aliases
- FAM39E, WASH
- Chromosome Location
- Chr. 9: 14475 - 30487 on Build GRCh38
- UniGene ID
- Hs.446466
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_182905.4 |
NP_878908.4
|
5, 6 |
683 |
|
XM_011517658.2 |
XP_011515960.1
|
5, 6 |
800 |
|
XM_011517659.2 |
XP_011515961.1
|
5, 6 |
582 |
|
XM_011517660.2 |
XP_011515962.1
|
5, 6 |
579 |
|
XM_011517662.2 |
XP_011515964.1
|
5, 6 |
2086 |
|
XM_011517663.2 |
XP_011515965.1
|
4, 5 |
1935 |
|
XM_011517664.2 |
XP_011515966.1
|
4, 5 |
1932 |
|
XM_011517665.2 |
XP_011515967.1
|
4, 5 |
481 |
|
XM_011517666.2 |
XP_011515968.1
|
4, 5 |
480 |
|
XM_017014169.1 |
XP_016869658.1
|
5, 6 |
699 |
|
XM_017014170.1 |
XP_016869659.1
|
5, 6 |
762 |
|
GenBank mRNA |
AK289708.1 |
- |
5, 6 |
686 |
|
- Gene Symbol
- WASH2P
- Entrez Gene ID
- 375260
- Gene Name
- WAS protein family homolog 2 pseudogene
- Gene Aliases
- FAM39B
- Chromosome Location
- Chr. 2: 113583653 - 113599037 on Build GRCh38
- UniGene ID
- Hs.446466
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY217347.1 |
- |
4, 5 |
1240 |
|
BC106020.1 |
- |
2, 3 |
232 |
|
BX537637.1 |
- |
5, 6 |
809 |
|
- Gene Symbol
- WASH3P
- Entrez Gene ID
- 374666
- Gene Name
- WAS protein family homolog 3 pseudogene
- Gene Aliases
- FAM39DP
- Chromosome Location
- Chr. 15: 101960813 - 101976605 on Build GRCh38
- UniGene ID
- Hs.459573
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC048328.1 |
- |
4, 5 |
557 |
|
- Gene Symbol
- WASH7P
- Entrez Gene ID
- 653635
- Gene Name
- WAS protein family homolog 7 pseudogene
- Gene Aliases
- FAM39F, WASH5P
- Chromosome Location
- Chr. 1: 14362 - 29370 on Build GRCh38
- UniGene ID
- Hs.446466
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK302089.1 |
- |
5, 6 |
636 |
|
View Full Product Details