Gene Transcripts

Gene Symbol
CLN8
Entrez Gene ID
2055
Gene Name
ceroid-lipofuscinosis, neuronal 8
Gene Aliases
C8orf61, EPMR
Chromosome Location
Chr. 8: 1755778 - 1786570 on Build GRCh38
UniGene ID
Hs.127675
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018941.3 NP_061764.2 3 5836
GenBank mRNA AK096736.1 - 1 530
BC136825.1 - 1 452 9020816

View Full Product Details