Gene Transcripts
- Gene Symbol
- FLNA
- Entrez Gene ID
- 2316
- Gene Name
- filamin A
- Gene Aliases
- ABP-280, ABPX, CSBS, CVD1, FLN, FLN-A, FLN1, FMD, MNS, NHBP, OPD, OPD1, OPD2, XLVD, XMVD
- Chromosome Location
- Chr. X: 154348529 - 154374638 on Build GRCh38
- UniGene ID
- Hs.195464
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK125630.1 |
- |
1 |
1147 |
|
View Full Product Details