Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001317812.1 | NP_001304741.1 | 4 | 3734 | |
NM_001317813.1 | NP_001304742.1 | 5 | 3381 | ||
NM_001317814.1 | NP_001304743.1 | 5 | 3328 | ||
NM_016612.3 | NP_057696.2 | 4 | 3257 | ||
GenBank mRNA | AF495725.1 | - | 1 | 1244 | |
BC042973.2 | - | 1 | 69 | ||
BX647289.1 | - | 1 | 370 |