Gene Transcripts

Gene Symbol
KLLN
Entrez Gene ID
100144748
Gene Name
killin, p53-regulated DNA replication inhibitor
Gene Aliases
CWS4, KILLIN
Chromosome Location
Chr. 10: 87859161 - 87863437 on Build GRCh38
UniGene ID
Hs.559820
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126049.1 NP_001119521.1 1 1209
GenBank mRNA EU552092.1 - 1 1209

View Full Product Details