Gene Transcripts

Gene Symbol
TRIM10
Entrez Gene ID
10107
Gene Name
tripartite motif containing 10
Gene Aliases
HERF1, RFB30, RNF9
Chromosome Location
Chr. 6: 30151939 - 30163216 on Build GRCh38
UniGene ID
Hs.709483
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006778.3 NP_006769.2 1 100
NM_052828.2 NP_439893.2 1 100
GenBank mRNA AF220122.1 - 1 95
AF220123.1 - 1 95
BC093926.1 - 1 78 7939771
BC113419.1 - 1 78 8322471

View Full Product Details