Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163278.1 | NP_001156750.1 | 28 | 5576 | |
NM_001163279.1 | NP_001156751.1 | 28 | 5573 | ||
NM_014253.3 | NP_055068.2 | 27 | 5555 | ||
XM_011531230.2 | XP_011529532.1 | 31 | 6217 | ||
XM_011531236.2 | XP_011529538.2 | 17 | 4794 | ||
XM_011531237.2 | XP_011529539.1 | 7 | 1678 | ||
XM_017029208.1 | XP_016884697.1 | 32 | 6316 | ||
XM_017029209.1 | XP_016884698.1 | 33 | 6371 | ||
XM_017029210.1 | XP_016884699.1 | 31 | 6244 | ||
XM_017029211.1 | XP_016884700.1 | 32 | 6322 | ||
XM_017029212.1 | XP_016884701.1 | 32 | 5933 | ||
XM_017029213.1 | XP_016884702.1 | 31 | 6245 | ||
XM_017029214.1 | XP_016884703.1 | 30 | 6196 | ||
GenBank mRNA | AF100772.1 | - | 27 | 5555 | |
BC140783.1 | - | 28 | 5573 | 9021651 | |
BC144543.1 | - | 28 | 5570 | 9053070 |