Gene Transcripts
- Gene Symbol
- MRVI1
- Entrez Gene ID
- 10335
- Gene Name
- murine retrovirus integration site 1 homolog
- Gene Aliases
- IRAG, JAW1L
- Chromosome Location
- Chr. 11: 10573091 - 10693988 on Build GRCh38
- UniGene ID
- Hs.501898
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001098579.2 |
NP_001092049.2
|
19, 20 |
2551 |
|
NM_001100163.2 |
NP_001093633.1
|
20, 21 |
2829 |
|
NM_001100167.2 |
NP_001093637.1
|
20, 21 |
2546 |
|
NM_001206880.1 |
NP_001193809.1
|
19, 20 |
2264 |
|
NM_001206881.1 |
NP_001193810.1
|
18, 19 |
2671 |
|
NM_130385.3 |
NP_569056.4
|
20, 21 |
2882 |
|
GenBank mRNA |
AF081249.1 |
- |
18, 19 |
2567 |
|
AF081250.1 |
- |
18, 19 |
2295 |
|
AK127209.1 |
- |
20, 21 |
2524 |
|
AK296336.1 |
- |
20, 21 |
2646 |
|
AK300358.1 |
- |
19, 20 |
2536 |
|
AK304702.1 |
- |
19, 20 |
1898 |
|
BC117127.1 |
- |
20, 21 |
2419 |
40125678 |
View Full Product Details