Gene Transcripts
- Gene Symbol
- CORO2B
- Entrez Gene ID
- 10391
- Gene Name
- coronin 2B
- Gene Aliases
- CLIPINC
- Chromosome Location
- Chr. 15: 68518373 - 68727806 on Build GRCh38
- UniGene ID
- Hs.551213
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001190456.1 |
NP_001177385.1
|
11, 12 |
1368 |
|
NM_001190457.1 |
NP_001177386.1
|
11, 12 |
1647 |
|
NM_001324014.1 |
NP_001310943.1
|
12, 13 |
1579 |
|
NM_001324015.1 |
NP_001310944.1
|
11, 12 |
1380 |
|
NM_006091.4 |
NP_006082.3
|
11, 12 |
1602 |
|
GenBank mRNA |
AB010098.1 |
- |
11, 12 |
1335 |
|
AB023142.1 |
- |
10, 11 |
1306 |
|
AK124711.1 |
- |
6, 7 |
1822 |
|
AK289678.1 |
- |
11, 12 |
1647 |
|
BC026335.1 |
- |
11, 12 |
1318 |
4792415 |
View Full Product Details