Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001098531.2 | NP_001092001.1 | 28 | 3253 | |
NM_001098532.2 | NP_001092002.1 | 27 | 3017 | ||
NM_006105.5 | NP_006096.2 | 28 | 2787 | ||
XM_005268571.3 | XP_005268628.1 | 27 | 2763 | ||
XM_011537752.1 | XP_011536054.1 | 28 | 3270 | ||
XM_011537755.1 | XP_011536057.1 | 28 | 2832 | ||
XM_011537758.2 | XP_011536060.1 | 28 | 2882 | ||
GenBank mRNA | AF103905.1 | - | 27 | 2577 | |
AK055502.1 | - | 10 | 1568 | ||
AK074444.1 | - | 12 | 1945 | ||
AK092448.1 | - | 28 | 3225 | ||
AK125545.1 | - | 24 | 2270 | ||
AK290230.1 | - | 29 | 2918 | ||
AK299029.1 | - | 27 | 2680 | ||
BC017728.1 | - | 28 | 2867 | 4414360 | |
BC092404.1 | - | 28 | 2856 | 6181000 | |
BQ007070.1 | - | 1 | 405 | ||
U78168.1 | - | 28 | 2792 |