Gene Transcripts
- Gene Symbol
- TDRKH
- Entrez Gene ID
- 11022
- Gene Name
- tudor and KH domain containing
- Gene Aliases
- TDRD2
- Chromosome Location
- Chr. 1: 151769354 - 151790576 on Build GRCh38
- UniGene ID
- Hs.144439
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001083963.1 |
NP_001077432.1
|
3, 4 |
382 |
|
NM_001083964.1 |
NP_001077433.1
|
3, 4 |
407 |
|
NM_001083965.1 |
NP_001077434.1
|
3, 4 |
407 |
|
NM_006862.3 |
NP_006853.2
|
3, 4 |
407 |
|
XM_005244856.4 |
XP_005244913.1
|
3, 4 |
273 |
|
XM_017000122.1 |
XP_016855611.1
|
3, 4 |
449 |
|
XM_017000123.1 |
XP_016855612.1
|
3, 4 |
449 |
|
XM_017000124.1 |
XP_016855613.1
|
3, 4 |
348 |
|
XM_017000125.1 |
XP_016855614.1
|
3, 4 |
377 |
|
XM_017000126.1 |
XP_016855615.1
|
3, 4 |
348 |
|
XM_017000127.1 |
XP_016855616.1
|
3, 4 |
377 |
|
XM_017000128.1 |
XP_016855617.1
|
3, 4 |
402 |
|
XM_017000129.1 |
XP_016855618.1
|
3, 4 |
367 |
|
XM_017000130.1 |
XP_016855619.1
|
3, 4 |
341 |
|
GenBank mRNA |
AF119121.2 |
- |
3, 4 |
395 |
|
AF227192.1 |
- |
3, 4 |
395 |
|
AK225160.1 |
- |
3, 4 |
361 |
|
AK226092.1 |
- |
3, 4 |
320 |
|
AK295948.1 |
- |
3, 4 |
278 |
|
AK303063.1 |
- |
3, 4 |
303 |
|
AK316279.1 |
- |
3, 4 |
358 |
|
BC032690.1 |
- |
3, 4 |
314 |
|
DA796638.1 |
- |
3, 4 |
407 |
|
View Full Product Details