Gene Transcripts
- Gene Symbol
- ADRM1
- Entrez Gene ID
- 11047
- Gene Name
- adhesion regulating molecule 1
- Gene Aliases
- ARM-1, ARM1, GP110
- Chromosome Location
- Chr. 20: 62302056 - 62308862 on Build GRCh38
- UniGene ID
- Hs.90107
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_007002.3 |
NP_008933.2
|
2, 3 |
364 |
|
NM_175573.2 |
NP_783163.1
|
2, 3 |
360 |
|
XM_005260257.1 |
XP_005260314.1
|
3, 4 |
451 |
|
XM_011528503.1 |
XP_011526805.1
|
2, 3 |
1199 |
|
XM_017027602.1 |
XP_016883091.1
|
2, 3 |
1199 |
|
XM_017027603.1 |
XP_016883092.1
|
3, 4 |
451 |
|
XM_017027604.1 |
XP_016883093.1
|
2, 3 |
1195 |
|
XM_017027605.1 |
XP_016883094.1
|
2, 3 |
1199 |
|
GenBank mRNA |
AK297567.1 |
- |
2, 3 |
258 |
|
AK307459.1 |
- |
|
294 |
|
AL558522.3 |
- |
2, 3 |
296 |
|
BC017245.2 |
- |
2, 3 |
260 |
4890786 |
D64154.1 |
- |
2, 3 |
255 |
|
HY029018.1 |
- |
|
457 |
|
View Full Product Details