Gene Transcripts

Gene Symbol
KLHL2
Entrez Gene ID
11275
Gene Name
kelch like family member 2
Gene Aliases
ABP-KELCH, MAV, MAYVEN
Chromosome Location
Chr. 4: 165207618 - 165323156 on Build GRCh38
UniGene ID
Hs.388668
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161521.1 NP_001154993.1 9 1107
NM_001161522.1 NP_001154994.1 8 1000
NM_007246.3 NP_009177.3 9 1230
XM_011531572.2 XP_011529874.1 10 1220
XM_011531575.2 XP_011529877.1 8 1052
XM_011531576.2 XP_011529878.1 7 720
XM_017007674.1 XP_016863163.1 10 1180
XM_017007675.1 XP_016863164.1 8 945
XM_017007676.1 XP_016863165.1 8 1053
XM_017007677.1 XP_016863166.1 7 927
GenBank mRNA AB209727.1 - 5 574
AF059569.1 - 9 1079
AK025289.1 - 6 696
AK294103.1 - 9 1101
AK294336.1 - 5 529
AK315372.1 - 9 1075
BC022503.1 - 9 1212 4791972
BC036468.2 - 9 1171 4799622
BX647776.1 - 8 984

View Full Product Details