Gene Transcripts

Gene Symbol
SFT2D1
Entrez Gene ID
113402
Gene Name
SFT2 domain containing 1
Gene Aliases
C6orf83, pRGR1
Chromosome Location
Chr. 6: 166319728 - 166342606 on Build GRCh38
UniGene ID
Hs.487143
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145169.2 NP_660152.1 1, 2 196
GenBank mRNA AF041429.1 - 1, 2 130
BC018969.1 - 1, 2 101 3959886
BG706927.1 - 1, 2 185

View Full Product Details