Gene Transcripts

Gene Symbol
SLC26A8
Entrez Gene ID
116369
Gene Name
solute carrier family 26 member 8
Gene Aliases
SPGF3, TAT1
Chromosome Location
Chr. 6: 35943514 - 36024855 on Build GRCh38
UniGene ID
Hs.435836
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193476.1 NP_001180405.1 20 2967
NM_052961.3 NP_443193.1 20 2991
NM_138718.2 NP_619732.2 18 2714
XM_011514294.2 XP_011512596.1 19 3132
XM_017010235.1 XP_016865724.1 20 3017
GenBank mRNA AF314959.1 - 19 2861
AF331522.1 - 20 2935
AI015633.1 - 1 442
AK055314.1 - 10 2437
AK057276.1 - 10 1842
AK097410.1 - 14 2053
AK122928.1 - 20 2967
BC025408.2 - 18 2707 4823098

View Full Product Details