Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001024647.3 | NP_001019818.1 | 4 | 579 | |
NM_001278402.1 | NP_001265331.1 | 4 | 494 | ||
NM_022456.4 | NP_071901.2 | 6 | 1230 | ||
NM_175623.3 | NP_783322.1 | 6 | 1364 | ||
NM_175624.3 | NP_783323.1 | 6 | 1230 | ||
NM_175625.3 | NP_783324.1 | 6 | 1364 | ||
NR_103519.1 | 6 | 1230 | |||
NR_103520.1 | 6 | 1364 | |||
XM_006719226.3 | XP_006719289.3 | 6 | 1353 | ||
XM_006719227.3 | XP_006719290.3 | 6 | 1026 | ||
XM_017018773.1 | XP_016874262.1 | 6 | 988 | ||
XM_017018774.1 | XP_016874263.1 | 8 | 1241 | ||
XM_017018776.1 | XP_016874265.1 | 7 | 1113 | ||
XM_017018777.1 | XP_016874266.1 | 7 | 1532 | ||
GenBank mRNA | AJ312896.1 | - | 6 | 1054 | |
AJ312897.1 | - | 6 | 1346 | ||
AJ312898.1 | - | 6 | 1054 | ||
AJ312899.1 | - | 6 | 1346 | ||
AK026201.1 | - | 6 | 1054 | ||
AK027566.1 | - | 4 | 490 | ||
AK027671.1 | - | 4 | 480 | ||
AK057844.1 | - | 6 | 959 | ||
AK225441.1 | - | 6 | 1054 | ||
AK314547.1 | - | 6 | 1065 | ||
BC015548.1 | - | 4 | 557 | 3846338 | |
BC059358.1 | - | 6 | 1019 | 5300636 | |
DN912041.1 | - | 4 | 579 |