Gene Transcripts

Gene Symbol
C16orf46
Entrez Gene ID
123775
Gene Name
chromosome 16 open reading frame 46
Gene Aliases
Chromosome Location
Chr. 16: 81053497 - 81077267 on Build GRCh38
UniGene ID
Hs.658684
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100873.1 NP_001094343.1 3 1210
NM_152337.2 NP_689550.2 4 1299
XM_005255792.2 XP_005255849.1 4 1247
XM_011522848.2 XP_011521150.2 4 1437
XM_011522849.2 XP_011521151.2 4 1466
XM_017022916.1 XP_016878405.1 4 1247
XM_017022917.1 XP_016878406.1 3 1197
GenBank mRNA AK057264.1 - 4 1231
AK295411.1 - 2 894
BC064143.1 - 3 1156 30389327

View Full Product Details