Gene Transcripts

Gene Symbol
CRYGC
Entrez Gene ID
1420
Gene Name
crystallin gamma C
Gene Aliases
CCL, CRYG3, CTRCT2
Chromosome Location
Chr. 2: 208128137 - 208139100 on Build GRCh38
UniGene ID
Hs.72910
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020989.3 NP_066269.1 3 440
XM_011510662.1 XP_011508964.1 4 514
GenBank mRNA BC074954.2 - 3 440
BC074955.2 - 3 440 30915501
BU737104.1 - 1 202
KT455014.1 - 3 440

View Full Product Details