Gene Transcripts

Gene Symbol
CDCA2
Entrez Gene ID
157313
Gene Name
cell division cycle associated 2
Gene Aliases
PPP1R81, Repo-Man
Chromosome Location
Chr. 8: 25458569 - 25511308 on Build GRCh38
UniGene ID
Hs.33366
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317906.1 NP_001304835.1 11, 12 2160
NM_001317907.1 NP_001304836.1 12, 13 2077
NM_152562.3 NP_689775.2 12, 13 2077
XM_006716294.2 XP_006716357.1 12, 13 2092
XM_011544415.1 XP_011542717.1 13, 14 2164
XM_011544416.2 XP_011542718.1 13, 14 1858
XM_011544417.2 XP_011542719.1 13, 14 1915
XM_011544418.1 XP_011542720.1 13, 14 2128
GenBank mRNA AK092164.1 - 7, 8 694
AK292505.1 - 11, 12 1892
AK299278.1 - 12, 13 1783
AL833396.1 - 9, 10 1182
AL833627.1 - 11, 12 1854
BC036214.1 - 9, 10 1205 5267276
BX649008.1 - 8, 9 1068

View Full Product Details