Gene Transcripts

Gene Symbol
AP1G1
Entrez Gene ID
164
Gene Name
adaptor related protein complex 1 gamma 1 subunit
Gene Aliases
ADTG, CLAPG1
Chromosome Location
Chr. 16: 71729000 - 71809073 on Build GRCh38
UniGene ID
Hs.461253
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001030007.1 NP_001025178.1 3, 4 635
NM_001128.5 NP_001119.3 3, 4 635
GenBank mRNA AB015317.1 - 3, 4 340
AK128078.1 - 6, 7 1190
AK225906.1 - 3, 4 394
AK294546.1 - 3, 4 394
AK299630.1 - 4, 5 569
AK314484.1 - 3, 4 640
BC036283.1 - 3, 4 600 4158263
BU902252.1 - 3, 4 635
Y12226.1 - 3, 4 349

View Full Product Details