Gene Transcripts

Gene Symbol
THAP5
Entrez Gene ID
168451
Gene Name
THAP domain containing 5
Gene Aliases
Chromosome Location
Chr. 7: 108562144 - 108569768 on Build GRCh38
UniGene ID
Hs.707689
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130475.2 NP_001123947.1 3 1294
NM_001287598.1 NP_001274527.1 3 1248
NM_001287599.1 NP_001274528.1 3 1172
NM_001287601.1 NP_001274530.1 2 1101
NM_182529.3 NP_872335.2 3 1370
GenBank mRNA AL833137.1 - 3 1249
BC053634.1 - 3 1370 5165075

Gene Symbol
THAP5P1
Entrez Gene ID
285872
Gene Name
THAP domain containing 5 pseudogene 1
Gene Aliases
Chromosome Location
Chr. 7: 158622173 - 158623381 on Build GRCh38
UniGene ID
Hs.490790
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK126609.1 - 1 2571

View Full Product Details