Gene Transcripts

Gene Symbol
TMEM268
Entrez Gene ID
203197
Gene Name
transmembrane protein 268
Gene Aliases
C9orf91
Chromosome Location
Chr. 9: 114603897 - 114646423 on Build GRCh38
UniGene ID
Hs.522357
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153045.3 NP_694590.2 8 1027
XM_006716997.1 XP_006717060.1 8 1030
XM_011518350.1 XP_011516652.1 9 1198
XM_011518351.2 XP_011516653.1 9 1326
XM_011518352.1 XP_011516654.1 9 1195
XM_011518353.1 XP_011516655.1 8 1087
XM_011518354.1 XP_011516656.1 8 1233
XM_011518355.1 XP_011516657.1 8 1230
XM_017014429.1 XP_016869918.1 9 1354
XM_017014430.1 XP_016869919.1 7 1119
GenBank mRNA AK095364.1 - 3 351
AK289383.1 - 8 996
AL834412.1 - 6 639
BC019913.1 - 2 200
BC062550.1 - 4 369
BC107133.1 - 8 965 40025594
BC107134.1 - 8 1178 40025600
BX649023.1 - 8 913

View Full Product Details