Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001433.3 | NP_001424.3 | 20, 21 | 2776 | |
XM_017024347.1 | XP_016879836.1 | 20, 21 | 4130 | ||
XM_017024348.1 | XP_016879837.1 | 19, 20 | 4028 | ||
GenBank mRNA | AB209869.1 | - | 20, 21 | 2759 | |
AF059198.1 | - | 20, 21 | 2759 | ||
AK292403.1 | - | 20, 21 | 2802 | ||
BC130405.1 | - | 20, 21 | 2709 | 40146436 | |
BC130407.1 | - | 20, 21 | 2709 | 40146438 |