Gene Transcripts

Gene Symbol
OR4D5
Entrez Gene ID
219875
Gene Name
olfactory receptor family 4 subfamily D member 5
Gene Aliases
OR11-276
Chromosome Location
Chr. 11: 123939617 - 123940573 on Build GRCh38
UniGene ID
Hs.553639
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001965.1 NP_001001965.1 1 412
GenBank mRNA BC136943.1 - 1 486
BC136945.1 - 1 486 9020937

View Full Product Details