Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001042729.1 | NP_001036194.1 | 7 | 782 | |
NM_001042747.1 | NP_001036212.1 | 7 | 841 | ||
NM_005248.2 | NP_005239.1 | 7 | 798 | ||
XM_006710452.2 | XP_006710515.1 | 8 | 849 | ||
XM_011541010.1 | XP_011539312.1 | 7 | 763 | ||
XM_011541011.2 | XP_011539313.1 | 5 | 562 | ||
XM_011541012.1 | XP_011539314.1 | 7 | 798 | ||
XM_011541013.2 | XP_011539315.1 | 7 | 798 | ||
XM_011541014.2 | XP_011539316.1 | 3 | 380 | ||
XM_017000673.1 | XP_016856162.1 | 5 | 418 | ||
XM_017000674.1 | XP_016856163.1 | 4 | 353 | ||
GenBank mRNA | AK130188.1 | - | 7 | 760 | |
AK301078.1 | - | 5 | 556 | ||
BC064382.1 | - | 7 | 711 | 5734966 | |
M19722.1 | - | 5 | 717 |