Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001193286.1 | NP_001180215.1 | 5 | 650 | |
NM_012237.3 | NP_036369.2 | 6 | 658 | ||
NM_030593.2 | NP_085096.1 | 5 | 650 | ||
NR_034146.1 | 4 | 581 | |||
XM_006723111.1 | XP_006723174.1 | 7 | 606 | ||
XM_011526654.1 | XP_011524956.1 | 5 | 543 | ||
XM_011526655.1 | XP_011524957.1 | 3 | 358 | ||
XM_017026500.1 | XP_016881989.1 | 5 | 414 | ||
GenBank mRNA | AB009274.1 | - | 5 | 374 | |
AF083107.2 | - | 6 | 556 | ||
AF095714.1 | - | 5 | 429 | ||
AF131800.1 | - | 4 | 305 | ||
AF160214.1 | - | 5 | 410 | ||
AJ505014.1 | - | 3 | 1127 | ||
AK025876.1 | - | 5 | 436 | ||
AK054642.1 | - | 6 | 489 | ||
AK131390.1 | - | 2 | 966 | ||
AK225824.1 | - | 5 | 433 | ||
AK290716.1 | - | 4 | 581 | ||
AK314492.1 | - | 6 | 412 | ||
AY030277.1 | - | 5 | 411 | ||
BC003012.1 | - | 5 | 501 | 2820929 | |
BC003547.1 | - | 5 | 501 | 3536728 | |
KF032391.1 | - | 3 | 358 |