Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001287059.1 | NP_001273988.1 | 14 | 3288 | |
NM_015052.4 | NP_055867.3 | 16 | 3625 | ||
XM_005249665.3 | XP_005249722.1 | 15 | 3407 | ||
XM_006715670.3 | XP_006715733.1 | 14 | 3305 | ||
XM_006715671.3 | XP_006715734.1 | 14 | 3302 | ||
XM_006715673.3 | XP_006715736.1 | 13 | 3200 | ||
XM_011515220.2 | XP_011513522.1 | 15 | 3198 | ||
XM_011515222.2 | XP_011513524.1 | 14 | 3339 | ||
XM_011515223.2 | XP_011513525.1 | 14 | 3140 | ||
XM_011515224.2 | XP_011513526.1 | 14 | 3620 | ||
XM_011515225.2 | XP_011513527.2 | 12 | 2657 | ||
XM_011515226.2 | XP_011513528.1 | 11 | 2881 | ||
XM_017011882.1 | XP_016867371.1 | 14 | 3647 | ||
XM_017011883.1 | XP_016867372.1 | 15 | 3584 | ||
XM_017011884.1 | XP_016867373.1 | 14 | 3644 | ||
XM_017011885.1 | XP_016867374.1 | 16 | 3580 | ||
XM_017011886.1 | XP_016867375.1 | 15 | 3409 | ||
XM_017011887.1 | XP_016867376.1 | 13 | 3545 | ||
XM_017011888.1 | XP_016867377.1 | 13 | 3542 | ||
XM_017011889.1 | XP_016867378.1 | 14 | 3599 | ||
XM_017011890.1 | XP_016867379.1 | 15 | 3409 | ||
GenBank mRNA | AB002320.2 | - | 16 | 3625 | |
AB048365.1 | - | 13 | 2977 | ||
AK294918.1 | - | 14 | 3242 | ||
BC151227.1 | - | 16 | 3625 | 9007236 |