Gene Transcripts
- Gene Symbol
- DPY19L1
- Entrez Gene ID
- 23333
- Gene Name
- dpy-19 like 1 (C. elegans)
- Gene Aliases
- Chromosome Location
- Chr. 7: 34928881 - 35038041 on Build GRCh38
- UniGene ID
- Hs.408623
- Species
- Homo sapiens
- Gene Symbol
- DPY19L1P2
- Entrez Gene ID
- 102724668
- Gene Name
- DPY19L1 pseudogene 2
- Gene Aliases
- THC1072510
- Chromosome Location
- Chr. 7: 32826302 - 32838744 on Build GRCh38
- UniGene ID
- Hs.636806
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_132360.1 |
|
6 |
943 |
|
GenBank mRNA |
DQ287935.1 |
- |
6 |
944 |
|
View Full Product Details