Gene Transcripts

Gene Symbol
ARFIP2
Entrez Gene ID
23647
Gene Name
ADP ribosylation factor interacting protein 2
Gene Aliases
POR1
Chromosome Location
Chr. 11: 6474683 - 6481479 on Build GRCh38
UniGene ID
Hs.732203
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242854.1 NP_001229783.1 4, 5 708
NM_001242855.1 NP_001229784.1 3, 4 468
NM_012402.3 NP_036534.1 4, 5 569
XM_005252840.4 XP_005252897.1 4, 5 473
GenBank mRNA AK026390.1 - 4, 5 401
AK301856.1 - 3, 4 423
AK301972.1 - 4, 5 510
AK311086.1 - 3, 4 1051
BC000392.2 - 4, 5 391 2821666
BM928413.1 - 536
U52522.1 - 4, 5 385
X97567.1 - 2, 3 204

View Full Product Details