Gene Transcripts
- Gene Symbol
- TXNDC8
- Entrez Gene ID
- 255220
- Gene Name
- thioredoxin domain containing 8
- Gene Aliases
- SPTRX-3, TRX6, bA427L11.2
- Chromosome Location
- Chr. 9: 110300900 - 110337887 on Build GRCh38
- UniGene ID
- Hs.147064
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001003936.3 |
NP_001003936.1
|
1, 2 |
101 |
|
NM_001286946.1 |
NP_001273875.1
|
1, 2 |
101 |
|
NM_001286947.1 |
NP_001273876.1
|
1, 2 |
101 |
|
XM_011518463.2 |
XP_011516765.1
|
1, 2 |
104 |
|
XM_017014579.1 |
XP_016870068.1
|
1, 2 |
104 |
|
XM_017014580.1 |
XP_016870069.1
|
1, 2 |
104 |
|
XM_017014581.1 |
XP_016870070.1
|
1, 2 |
104 |
|
XM_017014582.1 |
XP_016870071.1
|
1, 2 |
104 |
|
XM_017014583.1 |
XP_016870072.1
|
1, 2 |
104 |
|
GenBank mRNA |
AF305830.1 |
- |
1, 2 |
41 |
|
BC035743.1 |
- |
1, 2 |
62 |
|
BC130549.1 |
- |
1, 2 |
31 |
40146580 |
BC130551.1 |
- |
1, 2 |
31 |
40146582 |
BC144461.1 |
- |
1, 2 |
31 |
9052987 |
HY007335.1 |
- |
1, 2 |
102 |
|
View Full Product Details