Gene Transcripts

Gene Symbol
CHRDL2
Entrez Gene ID
25884
Gene Name
chordin like 2
Gene Aliases
BNF1, CHL2
Chromosome Location
Chr. 11: 74696428 - 74731385 on Build GRCh38
UniGene ID
Hs.432379
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278473.2 NP_001265402.1 8 1414
NM_001304390.1 NP_001291319.1 7 1306
NM_001304416.1 NP_001291345.1 6 1212
NM_001304417.1 NP_001291346.1 5 1004
NM_015424.5 NP_056239.3 8 1414
GenBank mRNA AF332891.1 - 5 695
AK304519.1 - 8 1182
AL110168.1 - 2 316
AY163868.1 - 8 920
AY279090.1 - 8 1211
AY279092.1 - 6 1212
AY279093.1 - 6 1212
AY279094.1 - 5 1118
AY358522.1 - 8 1203
BC142623.1 - 8 1414 40147467

View Full Product Details