Gene Transcripts

Gene Symbol
GALNS
Entrez Gene ID
2588
Gene Name
galactosamine (N-acetyl)-6-sulfatase
Gene Aliases
GALNAC6S, GAS, GalN6S, MPS4A
Chromosome Location
Chr. 16: 88813734 - 88856970 on Build GRCh38
UniGene ID
Hs.271383
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000512.4 NP_000503.1 2 280
NM_001323544.1 NP_001310473.1 3 454
XM_005256301.3 XP_005256358.1 2 280
XM_011522982.2 XP_011521284.1 3 326
XM_017023111.1 XP_016878600.1 2 2177
XM_017023112.1 XP_016878601.1 2 2177
GenBank mRNA AK131096.1 - 1 2865
AK308269.1 - 3 310
AK310392.1 - 3 326
BC050684.1 - 2 269 6091623
BC056151.1 - 2 261 6026248
DA010361.1 - 2 280
DA709138.1 - 2 284

View Full Product Details