Gene Transcripts

Gene Symbol
TOR1B
Entrez Gene ID
27348
Gene Name
torsin family 1 member B
Gene Aliases
DQ1
Chromosome Location
Chr. 9: 129803143 - 129811424 on Build GRCh38
UniGene ID
Hs.252682
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014506.2 NP_055321.1 5 856
NR_133948.1 5 997
NR_134027.1 3 414
GenBank mRNA AF007872.1 - 4 577
AF317129.1 - 5 846
BC015578.2 - 5 801 4589479
CN262366.1 - 4 646
HY036797.1 - 3 374

View Full Product Details