Gene Transcripts

Gene Symbol
SLC25A47
Entrez Gene ID
283600
Gene Name
solute carrier family 25 member 47
Gene Aliases
C14orf68, HDMCP, HMFN1655
Chromosome Location
Chr. 14: 100323342 - 100330378 on Build GRCh38
UniGene ID
Hs.108268
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207117.2 NP_997000.2 6 937
GenBank mRNA AB073385.1 - 7 861
AK092001.1 - 6 941
AY569438.1 - 6 937
BC137253.1 - 6 917 9021250
BC137254.1 - 6 917 9021251
BC144549.1 - 6 978
BX248260.1 - 6 926

View Full Product Details