Gene Transcripts
- Gene Symbol
- HEXDC
- Entrez Gene ID
- 284004
- Gene Name
- hexosaminidase D
- Gene Aliases
- Chromosome Location
- Chr. 17: 82418376 - 82442645 on Build GRCh38
- UniGene ID
- Hs.744207
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_173620.2 |
NP_775891.2
|
5, 6 |
856 |
|
XM_005256346.3 |
XP_005256403.1
|
4, 5 |
444 |
|
XM_005256347.4 |
XP_005256404.1
|
5, 6 |
580 |
|
XM_006722280.3 |
XP_006722343.1
|
4, 5 |
419 |
|
XM_011523560.2 |
XP_011521862.1
|
3, 4 |
387 |
|
XM_011523561.2 |
XP_011521863.1
|
4, 5 |
425 |
|
XM_017024484.1 |
XP_016879973.1
|
5, 6 |
580 |
|
XM_017024485.1 |
XP_016879974.1
|
4, 5 |
444 |
|
GenBank mRNA |
AK074405.1 |
- |
4, 5 |
419 |
|
BC018205.1 |
- |
4, 5 |
451 |
3464710 |
BC035561.1 |
- |
5, 6 |
537 |
5211583 |
FM204887.1 |
- |
4, 5 |
440 |
|
View Full Product Details